Learn More
Quickfit™ Borosilicate Glass Medium Neck Flat Bottom Flask
Marke: Quickfit FF500/3UM
Weitere Details : Netto-Gewicht : 0.03000kg
Beschreibung
- With longer neck than standard Quickfit™ flasks
- Useful in reducing the problems associated with splashing and frothing