missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Protein Fraction II Protein Fraction
N.V.
Spezifikation
Konjugat | Unconjugated |
---|---|
Zusammensetzung | Supplied as a solution in HEPES. |
Name | Protein Fraction II |
Beschreibung
Source: Human cervical epithelial carcinoma cell line, HeLa-derived
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO
Spezifikation
Unconjugated | |
Supplied as a solution in HEPES. | |
Protein Fraction II |