Learn More
R&D Systems™ MagCellect CD24- CD44+ Breast Cancer Stem Cell Isolation Kit
Artikelnummer. 18422490
Change view
Click to view available options
Menge:
1 Kit
Packungsgröße:
1 Set
Artikelnummer. Menge unitSize
18422490 1 Kit 1 Set
1 options
Artikelnummer. 18422490
Marke: R&D Systems™ MAGH111
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Isolation by positive and negative selection improves purity of recovered cells
Cancer stem cells are a rare population of self-renewing cells found in tumors and in cancer cell lines. Methods that use positive or negative selection to enrich for cancer stem cells can lead to poor recovery and low cell purity as well as the loss of valuable tissue samples and resources. Methods that use both positive and negative selection to isolate cancer stem cells can yield cells with a higher purity.
Isolation based on two markers increases the purity of recovered cells
Isolates CD24low/- CD44+ cells which have been shown to form tumors in mice
Includes fluorochrome-conjugated CD24 and CD44 antibodies for analysis of recovered cells
TRUSTED_SUSTAINABILITY
Spezifikation
Inhalt und Lagerung Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Menge 1 Kit