missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 40-113)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

N.V.

Spezifikation

Gen-ID (Entrez) 57349
Molekulargewicht 8.2 kDa
Name CXCL7/Thymus Chemokine-1 aa 40-113
Menge 25μg
Quelle E. coli-derived mouse CXCL7/Thymus Chemokine-1 protein Lys40-Tyr113
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 40-113) is derived from E. coli. The Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 40-113) has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

57349
CXCL7/Thymus Chemokine-1 aa 40-113
E. coli-derived mouse CXCL7/Thymus Chemokine-1 protein Lys40-Tyr113
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
8.2 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen