missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Donkey IgG (H+L) Secondary Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen PA128591
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-28591 detects Donkey IgG from donkey samples.
Donkey IgG (H+L) Polyclonal antibody specifically detects Donkey IgG (H+L) in Donkey samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Donkey IgG (H+L) | |
Polyclonal | |
Biotin | |
Rabbit | |
IgG fraction | |
RUO | |
Donkey | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
2.0 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
donkey IgG whole molecule | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |