Learn More
Rabbit anti-Human IgG Fc Secondary Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen 31789
Weitere Details : Netto-Gewicht : 0.00150kg
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31789 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31789 reacts with the heavy chains of human IgG, but not with the light chains common to most human immunoglobulins. The antibody does not react against human IgM or IgA, or against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.5 mL of distilled water. Country of Origin: USA
Human IgG Fc Polyclonal antibody specifically detects Human IgG Fc in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Human IgG Fc | |
Polyclonal | |
Biotin | |
Rabbit | |
1.5 mL | |
Secondary | |
4° C | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.2 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Human | |
Lyophilized |