Learn More
Rabbit anti-Human IgM (Heavy chain) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen A18911
175.75 EUR gültig bis 2024-05-31
Benutzen Sie den Promotions-Code "21664" um Ihren Promotions-Preis zu erhalten.
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Human IgM (Heavy chain) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity chromatography | |
RUO | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Human Mu immunoglobin | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |