Learn More
ACROBiosystems Rabbit Complement C5 Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | XP_008271599.1 |
---|---|
Molekulargewicht | 73.8 kDa (β chain) and 111.9 kDa (α chain) |
Quelle | Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession # XP_008271599.1). |
Immunogen | Gln 19 - Ala 1678 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16447439
|
ACROBiosystems
CO5-R52H4-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 05-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16457439
|
ACROBiosystems
CO5-R52H4-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Derived from proteolytic degradation of complement C5, C5 anaphylatoxin is a mediator of local inflammatory process. C5 precursor is first processed by the removal of 4 basic residues, forming two chains, beta and alpha, linked by a disulfide bond. C5 convertase activates C5 by cleaving the alpha chain, releasing C5a anaphylatoxin and generating C5b (beta chain + alpha' chain). Activation of C5 by a C5 convertase initiates the spontaneous assembly of the late complement components, C5-C9, into the membrane attack complex. C5b has a transient binding site for C6. The C5b-C6 complex is the foundation upon which the lytic complex is assembled. The C5a anaphylatoxin interacts with C5AR1 and tick complement inhibitor. C5a is also a potent chemokine which stimulates the locomotion of polymorphonuclear leukocytes and directs their migration toward sites of inflammation.
This protein carries a polyhistidine tag at the C-terminus. The mature form of Complement C5 is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. During cultivation, Complement C5 will be partially processed into α and β cSpezifikation
XP_008271599.1 | |
Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession # XP_008271599.1). | |
-20°C | |
Complement C5 | |
Rabbit | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
73.8 kDa (β chain) and 111.9 kDa (α chain) | |
Gln 19 - Ala 1678 | |
Complement C5,C5,CPAMD4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |