missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Radixin Recombinant Rabbit Monoclonal Antibody (24GB1420)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557674
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Radixin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Radixin is a cytoskeletal protein that may be important in linking actin to the plasma membrane. It is highly similar in sequence to both ezrin and moesin. The radixin gene has been localized by fluorescence in situ hybridization to 11q23. A truncated version representing a pseudogene (RDXP2) was assigned to Xp21.3. Another pseudogene that seemed to lack introns (RDXP1) was mapped to 11p by Southern and PCR analyses. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2012].
Spezifikation
| Radixin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Rdx | |
| AA516625; DFNB24; ESP10; radixin; radixin isoform; Rdx | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19684, 315655, 5962 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1420 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26043, P35241 | |
| Rdx | |
| A synthesized peptide derived from human Radixin (aa 350-430). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur