missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ran Antibody (ARAN1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Marke: Novus Biologicals NB600-1082-0.025ml
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ran Monoclonal specifically detects Ran in Human, Mouse, Bovine, Xenopus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Spezifikation
| Ran | |
| Monoclonal | |
| 2.5 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| RAN | |
| Denatured recombinant protein (Human). | |
| 0.025 mL | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 5901 | |
| Human | |
| Purified |
| Western Blot | |
| ARAN1 | |
| Unconjugated | |
| Androgen receptor-associated protein 24, ARA24Gsp1, GTPase Ran, GTP-binding nuclear protein Ran, guanosine triphosphatase Ran, member RAS oncogene family, OK/SW-cl.81, RAN, member RAS oncogene family, RanGTPase, Ras-like protein TC4, Ras-related nuclear protein, TC4 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Ran (ARAN1) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur