missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10469-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).Spezifikation
| RAN | |
| Polyclonal | |
| Unconjugated | |
| RAN | |
| ARA24, Gsp1, GTPase Ran, RAN, Ras like protein TC4, Ras related nuclear protein, TC4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19384, 5901, 84509 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62826, P62827, P62828 | |
| RAN | |
| RAN Fusion Protein Ag0748 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur