Learn More
RANGAP1 Mouse anti-Human, Unconjugated, Clone: 1F3A4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 67146-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The RANGAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RANGAP1. This antibody recognizes human, mouse, rat antigen. The RANGAP1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Spezifikation
| RANGAP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 1F3A4 | |
| Unconjugated | |
| 150 μL | |
| 5905 | |
| IgG1 |