Learn More
Invitrogen™ RARB Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Retinoic acid receptor beta, ligand binding domain (RARB-LBD) is a 59.20 kDa recombinant human protein (amino acids 168-448) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Marke: Invitrogen™ PV4716
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
RARB-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Spezifikation
Nuclear Receptor Assay | |
Store in ultra-cold freezer (-68 to -85°C). | |
Ligand Binding Domain | |
Human | |
Hormone Receptors |
50 μg | |
RUO - research use only | |
RAR-Beta LBD | |
Recombinant |