Learn More
CD115 Rat anti-Mouse, PE, Clone: T38-320, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 565249
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The T38-320 monoclonal antibody specifically binds to CD115 which is also known as Colony stimulating factor 1 Receptor (CSF-1R) or Macrophage colony-stimulating factor 1 receptor (M-CSFR). This type I transmembrane glycoprotein is a receptor tyrosine kinase (RTK) that belongs to the Ig superfamily. It is expressed on a variety of cells including those committed to the mononuclear phagocyte lineage, such as, monocytes, macrophages, and osteoclasts. CSF-1 binds to and signals through CSF-1R homodimers which undergo tyrosine autophosphorylation and transduce downstream signaling pathways resulting in cytoskeletal reorganization and gene expression. CSF-1R activation stimulates the proliferation, differentiation, and survival of cells within the mononuclear phagocyte system. Interleukin-34 (IL-34) is another ligand for CD115 that can induce similar, as well as, some different biological responses by CD115-positive target cells.
Spezifikation
CD115 | |
Monoclonal | |
0.2mg/mL | |
M-CSFR; M-CSF-R; CSF-1 Receptor; CSF-1R; Csf1r; Csfmr; Fms; c-Fms; Fim-2 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
T38-320 | |
PE | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Mouse CD115 Recombinant Protein | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |