Learn More
R&D Systems™ Rat CLEC10A/CD301 (NP_071788) VersaClone cDNA
Macrophage Galactose N-Acetyl-Galactosamine Specific Lectin 1
Marke: R&D Systems™ RDC1927
Beschreibung
The Rat CLEC10A/CD301 (NP_071788) VersaClone cDNA clone contains the complete ORF for CLEC10A/CD301, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CLEC10A/CD301 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Spezifikation
| rClec10a | |
| 10 μg at 0.2 μg/μL | |
| 934bp | |
| Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| 10 μg |
| NP_071788 | |
| cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
| 10462 (Human) | |
| Rat |