Learn More
RDM1 Mouse anti-Human, Unconjugated, Clone: 1C10B4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66941-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The RDM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RDM1. This antibody recognizes human, mouse, rat antigen. The RDM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| RDM1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1C10B4 | |
| Unconjugated | |
| 150 μL | |
| 201299 | |
| IgG2a |