Learn More
DiscoverX Ready-to-Assay™ C5aR Anaphylotoxin Receptor Frozen Cells
Beschreibung
The Ready-to-Assay™ C5aR Anaphylotoxin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. C5a) based activation of C5AR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress C5AR1 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of C5AR1 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Spezifikation
Spezifikation
| Assay-Eingabe | C5aR |
| Zur Verwendung mit (Anwendung) | Frozen Cells |
| Primärer Agonist | C5a |
| Menge | 2 Fläschchen mit Zellen |
| Zieleintrag | C5AR1 |