Learn More
enQuireBio™ Recombinant Chikungunya Virus VLP Protein
Recombinant Protein
Marke:  enQuireBio™ QP10473-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15903988
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.
Spezifikation
Spezifikation
VLP Protein
Research Use Only
Recombinant Protein
Human
0.02M tris-HCl pH7.5, 0.125M sodium chloride.
100 μg
Untested
Chikungunya virus (Strain: Senegal/37997)
Native
>90% as determined by SDS-PAGE.