missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia W3-W6 Protein
A cDNA sequence encoding the Chlamydia W3-W6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11415-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
W3-W6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl, pH 6.5, 100mM Sodium Phosphate and 8M urea. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged | |
Chlamydia W3-W6 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |