missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 8-oxo-dGTP diphosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) 8-oxo-dGTP diphosphatase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7301-ec-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_414641.1 | |
8-oxo-dGTP diphosphatase Protein | |
Research Use Only | |
mutT | |
Recombinant Protein | |
E. coli | |
MKKLQIAVGI IRNENNEIFI TRRAADAHMA NKLEFPGGKI EMGETPEQAV VRELQEEVGI TPQHFSLFEK LEYEFPDRHI TLWFWLVERW EGEPWGKEGQ PGEWMSLVGL NADDFPPANE PVIAKLKRL | |
Greater than 90% as determined by SDS-PAGE. |
944824 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |