missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L18 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L18, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8388-ec-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_000970.1 | |
60S ribosomal protein L18 Protein | |
Research Use Only | |
RPL18 | |
Recombinant Protein | |
E. coli | |
GVDIRHNKDR KVRRKEPKSQ DIYLRLLVKL YRFLARRTNS TFNQVVLKRL FMSRTNRPPL SLSRMIRKMK LPGRENKTAV VVGTITDDVR VQEVPKLKVC ALRVTSRARS RILRAGGKIL TFDQLALDSP KGCGTVLLSG PRKGREVYRH FGKAPGTPHS HTKPYVRSKG RKFERARGRR ASRGYK | |
Greater than 90% as determined by SDS-PAGE. |
6141 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |