missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Acidic mammalian chitinase Protein
A DNA sequence encoding the Homo sapiens (Human) Acidic mammalian chitinase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8133-ec-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001035713.1 | |
Acidic mammalian chitinase Protein | |
Research Use Only | |
CHIA | |
Recombinant Protein | |
E. coli | |
MVSTPENRQT FITSVIKFLR QYEFDGLDFD WEYPGSRGSP PQDKHLFTVL VQEMREAFEQ EAKQINKPRL MVTAAVAAGI SNIQSGYEIP QLSQYLDYIH VMTYDLHGSW EGYTGENSPL YKYPTDTGSN AYLNVDYVMN YWKDNGAPAE KLIVGFPTYG HNFILSNPSN TGIGAPTSGA GPAGPYAKES GIWAYYEICT FLKNGATQGW DAPQEVPYAY QGNVWVGYDN IKSFDIKAQW LKHNKFGGAM VWAIDLDDFT GTFCNQGKFP LISTLKKALG LQSASCTAPA QPIEPITAAP SGSGNGSGSS SSGGSSGGSG FCAVRANGLY PVANNRNAFW HCVNGVTYQQ NCQAGLVFDT SCDCCNWA | |
Greater than 80% as determined by SDS-PAGE. |
27159 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |