missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AMBP Protein
A cDNA sequence encoding the AMBP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11009-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
AMBP Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MRSLGALLLL LSACLAVSAG PVPTPPDNIQ VQENFNISRI YGKWYNLAIG STCPWLKKIM DRMTVSTLVL GEGATEAEIS MTSTRWRKGV CEETSGAYEK TDTDGKFLYH KSKWNITMES YVVHTNYDEY AIFLTKKFSR HHGPTITAKL YGRAPQLRET LLQDFRVVAQ GVGIPEDSIF TMADRGECVP GEQEPEPILI PRVRRAVLPQ EEEGSGGGQL VTEVTKKEDS CQLGYSAGPC MGMTSRYFYN GTSMACETFQ YGGCMGNGNN FVTEKECLQT CRTVAACNLP IVRGPCRAFI QLWAFDAVKG KCVLFPYGGC QGNGNKFYSE KECREYCGVP GDGDEELLRF SN | |
Greater than 96.0%. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Lyophilized from 0.02M NH4HCO3. |