Learn More
R&D Systems™ Recombinant Human Angiogenin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke: R&D Systems™ 265-AN-050
Weitere Details : Netto-Gewicht : 0.00970kg
Beschreibung
The Recombinant Human Angiogenin Protein is derived from E. coli. The Recombinant Human Angiogenin Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
14 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
283 | |
Angiogenin | |
E. coli-derived human Angiogenin protein Gln25-Pro147 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |