missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA4 Protein
A cDNA sequence encoding the ANXA4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11034-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
ANXA4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAMATKGGTV KAASGFNAME DAQTLRKAMK GLGTDEDAII SVLAYRNTAQ RQEIRTAYKS TIGRDLIDDL KSELSGNFEQ VIVGMMTPTV LYDVQELRRA MKGAGTDEGC LIEILASRTP EEIRRISQTY QQQYGRSLED DIRSDTSFMF QRVLVSLSAG GRDEGNYLDD ALVRQDAQDL YEAGEKKWGT DEVKFLTVLC SRNRNHLLHV FDEYKRISQK DIEQSIKSET SGSFEDALLA IVKCMRNKSA YFAEKLYKSM KGLGTDDNTL IRVMVSRAEI DMLDIRAHFK RLYGKSLYSF IKGDTSGDYR KVLLVLCGGD D | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The ANXA4 protein solution contains 20mM Tris-HCl, pH 8 and 20% glycerol, 0.2M NaCl. |