missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCAR1 Protein
A cDNA sequence encoding the BCAR1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11133-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
BCAR1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRLQQGVS ATVAHLLDLA GSAGATGSWR SPSEPQEPLV QDLQAAVAAV QSAVHELLEF ARSAVGNAAH TSDRALHAKL SRQLQKMEDV HQTLVAHGQA LDAGRGGSGA TLEDLDRLVA CSRAVPEDAK QLASFLHGNA SLLFRRTKAT APGPEGGGTL HPNPTDKTSS IQSRPLPSPP KFTSQDSPDG QYENSEGGWM EDYDYVHLQG KEEFEKTQKE LLEKGSITRQ GKSQLELQQL KQFERLEQEV SRPIDHDLAN WTPAQPLAPG RTGGLGPSDR QLLLFYLEQC EANLTTLTNA VDAFFTAVAT NQPPKIFVAH SKFVILSAHK LVFIGDTLSR QAKAADVRSQ VTHYSNLLCD LLRGIVATTK AAALQYPSPS AAQDMVE | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The BCAR1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |