missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5880-ye-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001276841.1 | |
Casein kinase I isoform epsilon Protein | |
Research Use Only | |
CSNK1E | |
Recombinant Protein | |
Yeast | |
MELRVGNKYR LGRKIGSGSF GDIYLGANIA SGEEVAIKLE CVKTKHPQLH IESKFYKMMQ GGVGIPSIKW CGAEGDYNVM VMELLGPSLE DLFNFCSRKF SLKTVLLLAD QMISRIEYIH SKNFIHRDVK PDNFLMGLGK KGNLVYIIDF GLAKKYRDAR THQHIPYREN KNLTGTARYA SINTHLGIEQ SRRDDLESLG YVLMYFNLGS LPWQGLKAAT KRQKYERISE KKMSTPIEVL CKGYPSEFST YLNFCRSLRF DDKPDYSYLR QLFRNLFHRQ GFSYDYVFDW NMLKFGAARN PEDVDRERRE HEREERMGQL RGSATRALPP GPPTGATANR LRSAAEPVAS TPASRIQPAG NTSPRAISRV DRERKVSMRL HRGAPANVSS SDLTGRQEVS RIPASQTSVP FDHLGK | |
Greater than 90% as determined by SDS-PAGE. |
102800317 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |