missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrosomal protein of 83 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 83 kDa, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8282-ec-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_057206.2 | |
Centrosomal protein of 83 kDa Protein | |
Research Use Only | |
CCDC41 | |
Recombinant Protein | |
E. coli | |
MLIDERLRCE HHKANYQTLK AEHTRLQNEH VKLQNELKHL FNEKQTQQEK LQLLLEELRG ELVEKTKDLE EMKLQILTPQ KLELLRAQIQ QELETPMRER FRNLDEEVEK YRAVYNKLRY EHTFLKSEFE HQKEEYARIL DEGKIKYESE IARLEEDKEE LRNQLLNVDL TKDSKRVEQL AREKVYLCQK LKGLEAEVAE LKAEKENSEA QVENAQRIQV RQLAEMQATV RSLEAEKQSA NLRAERLEKE LQSSSEQNTF LINKLHKAER EINTLSSKVK ELKHSNKLEI TDIKLETARA KSELERERNK IQSELDGLQS DNEILKAAVE HHKVLLVEKD RELIRKVQAA KEEGYQKLVV LQDEKLELEN RLADLEKMKV EHDVWRQSEK DQYEEKLRAS QMAEEITRKE LQSVRLKLQQ QIVTIENAEK EKNENSDLKQ QISSLQIQVT SLAQSENDLL NSNQMLKEMV ERLKQECRNF RSQAEKAQLE AEKTLEEKQI QWLEEKHKLH ERITDREEKY NQAKEKLQRA AIAQKKLEQD LELGCPSVTD TYRESVFPPP PLKRDLLK | |
Greater than 90% as determined by SDS-PAGE. |
51134 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |