missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLDN4 Protein
A cDNA sequence encoding the CLDN4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11441-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CLDN4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS MWRVTAFIGS NIVTSQTIWE GLWMNCVVQS TGQMQCKVYD SLLALPQDLQ AAR LEVLFQG PTAHNIIQDF YNPLVASGQK REM MWRVTAFIGS NIVTSQTIWE GLWMNCVVQS TGQMQCKVYD SLLALPQDLQ AAR LEVLFQG PTAHNIIQDF YNPLVASGQK REM | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |