missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNPY2 Protein
A DNA sequence encoding the Homo sapiens (Human) CNPY2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8489-ec-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001177920.1 | |
CNPY2 Protein | |
Research Use Only | |
CNPY2 | |
Recombinant Protein | |
E. coli | |
RSQDLHCGAC RALVDELEWE IAQVDPKKTI QMGSFRINPD GSQSVVEVPY ARSEAHLTEL LEEICDRMKE YGEQIDPSTH RKNYVRVVGR NGESSELDLQ GIRIDSDISG TLKFACESIV EEYEDELIEF FSREADNVKD KLCSKRTDLC DHALHISHDE L | |
Greater than 90% as determined by SDS-PAGE. |
10330 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |