Learn More
DiscoverX Recombinant Human Gas6
Artikelnummer. 17322639
Encompass
Change view
Click to view available options
:
500 μg
Artikelnummer.
17322639 500 μg
1 options
Artikelnummer. 17322639
Marke: DiscoverX 921326
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Human Gas6 is an agonist for AXL, TYRO3, and MERTK. Provided as 50μg of dry powder alongside reconstitution buffer.
Gas6 (Growth Arrest Specific 6) is a secreted, vitamin K dependent protein involved in several biological responses such as stimulation of cell proliferation, prevention of apoptosis, and differentiation and expansion of NK cell precursors. Acts as an agonist for AXL, TYRO3, and MERTK. Provided as 50μg of dry powder alongside reconstitution buffer.
Spezifikation
Menge 50 μg
Produkttyp Control Ligands