missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMOX2 Protein
A cDNA sequence encoding the HMOX2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12287-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
HMOX2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SAEVETSEG VDESEKKNSG ALEKENQMRM ADLSELLKEG TKEAHDRAEN TQFVKDFLKG NIKKELFKLA TTALYFTYSA LEEEMERNKD HPAFAPLYFP MELHRKEALT KDMEYFFGEN WEEQVQCPKA AQKYVERIHY IGQNEPELLV AHAYTRYMGD LSGGQVLKKV AQRALKLPST GEGTQFYLFE NVDNAQQFKQ LYRARMNALD LNMKTKERIV EEANKAFEYN MQIFNELDQA GSTLARETLE DGFPVHDGKG DMRK | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
HMOX2 solution containing 20mM Tris pH 8.0, 1mM DTT and 10% glycerol. |