missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MED4 Protein
A cDNA sequence encoding the MED4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12673-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
MED4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAASSSGEKE KERLGGGLGV AGGNSTRERL LSALEDLEVL SRELIEMLAI SRNQKLLQAG EENQVLELLI HRDGEFQELM KLALNQGKIH HEMQVLEKEV EKRDGDIQQL QKQLKEAEQI LATAVYQAKE KLKSIEKARK GAISSEEIIK YAHRISASNA VCAPLTWVPG DPRRPYPTDL EMRSGLLGQM NNPSTNGVNG HLPGDALAAG RLPDVLAPQY PWQSNDMSMN MLPPNHSSDF LLEPPGHNKE DEDDVEIMST DSSSSSSESD LEHHHHHH | |
Greater than 80.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MED4 protein solution contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 1mM DTT and 100mM NaCl. |