missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP1 Protein
A DNA sequence encoding the Homo sapiens (Human) MMP1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8564-ec-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_002412.1 | |
MMP1 Protein | |
Research Use Only | |
MMP1 | |
Recombinant Protein | |
E. coli | |
VLTEGNPRWE QTHLTYRIEN YTPDLPRADV DHAIEKAFQL WSNVTPLTFT KVSEGQADIM ISFVRGDHRD NSPFDGPGGN LAHAFQPGPG IGGDAHFDED ERWTNNFREY NLHRVAAHEL GHSLGLSHST DIGALMYPSY TFSGDVQLAQ DDIDGIQAIY GRSQNPVQPI GPQTPKACDS KLTFDAITTI RGEVMFFKDR FYMRTNPFYP EVELNFISVF WPQLPNGLEA AYEFADRDEV RFFKGNKYWA VQGQNVLHGY PKDIYSSFGF PRTVKHIDAA LSEENTGKTY FFVANKYWRY DEYKRSMDPG YPKMIAHDFP GIGHKVDAVF MKDGFFYFFH GTRQYKFDPK TKRILTLQKA NSWFNCRKN | |
Greater than 90% as determined by SDS-PAGE. |
4312 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |