missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPL48 Protein
A cDNA sequence encoding the MRPL48 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12731-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
MRPL48 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSGEKPIY SVGGILLSIS RPYKTKPTHG IGKYKHLIKA EEPKKKKGKV EVRAINLGTD YEYGVLNIHL TAYDMTLAES YAQYVHNLCN SLSIKVEESY AMPTKTIEVL QLQDQGSKML LDSVLTTHER VVQISGLSAT FAEIFLEIIQ SSLPEGVRLS VKEHTEEDFK GRFKARPELE ELLAKLK | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPL48 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |