missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS28 Protein
A cDNA sequence encoding the MRPS28 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12735-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
MRPS28 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGSPKNVE SFASMLRHSP LTQMGPAKDK LVIGRIFHIV ENDLYIDFGG KFHCVCRRPE VDGEKYQKGT RVRLRLLDLE LTSRFLGATT DTTVLEANAV LLGIQESKDS RSKEEHHEK | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPS28 protein solution (0.25 mg/ml) containing 20mM Phosphate buffer saline (pH 8.0), 20% glycerol and 1mM DTT. |