Learn More
enQuireBio™ Recombinant Human Myelin expression factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Myelin expression factor 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6399-ec-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001288139.1 | |
Myelin expression factor 2 Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
50804 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MYEF2 | |
Recombinant Protein | |
E. coli | |
GGMNRIGGGI GFGGLEAMNS MGGFGGVGRM GELYRGAMTS SMERDFGRGD IGINQGFGDS FGRLGSAMIG GFAGRIGSSN MGPVGSGISG GMGSMNSVTG GMGMGLDRMS SSFDRMGPGI GAILERSIDM DRGFLSGPMG SGMRERIGSK GNQIFVRNLP FDLTWQKLKE KFSQCGHVMF AEIKMENGKS KGCGTVRFDS PESAE |