missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin light chain 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin light chain 3, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6401-ec-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_000249.1 | |
Myosin light chain 3 Protein | |
Research Use Only | |
MYL3 | |
Recombinant Protein | |
E. coli | |
APKKPEPKKD DAKAAPKAAP APAPPPEPER PKEVEFDASK IKIEFTPEQI EEFKEAFMLF DRTPKCEMKI TYGQCGDVLR ALGQNPTQAE VLRVLGKPRQ EELNTKMMDF ETFLPMLQHI SKNKDTGTYE DFVEGLRVFD KEGNGTVMGA ELRHVLATLG ERLTEDEVEK LMAGQEDSNG CINYEAFVKH IMSS | |
Greater than 80% as determined by SDS-PAGE. |
4634 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |