missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NGB Protein
A cDNA sequence encoding the NGB was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12840-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NGB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MERPEPELIR QSWRAVSRSP LEHGTVLFAR LFALEPDLLP LFQYNCRQFS SPEDCLSSPE FLDHIRKVML VIDAAVTNVE DLSSLEEYLA SLGRKHRAVG VKLSSFSTVG ESLLYMLEKC LGPAFTPATR AAWSQLYGAV VQAMSRGWDG E | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Filtered and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer, 0.1M NaCl, pH 7.2. |