missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT5 Protein
A cDNA sequence encoding the NUDT5 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12907-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NUDT5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MESQEPTESS QNGKQYIISE ELISEGKWVK LEKTTYMDPT GKTRTWESVK RTTRKEQTAD GVAVIPVLQR TLHYECIVLV KQFRPPMGGY CIEFPAGLID DGETPEAAAL RELEEETGYK GDIAECSPAV CMDPGLSNCT IHIVTVTING DDAENARPKP KPGDGEFVEV ISLPKNDLLQ RLDALVAEEH LTVDARVYSY ALALKHANAK PFEVPFLKF | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUDT5 Human 1 mg/ml solution containing 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 10% glycerol. |