missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PNMT Protein
A cDNA sequence encoding the PNMT was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13083-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
PNMT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSGADRSPNA GAAPDSAPGQ AAVASAYQRF EPRAYLRNNY APPRGDLCNP NGVGPWKLRC LAQTFATGEV SGRTLIDIGS GPTVYQLLSA CSHFEDITMT DFLEVNRQEL GRWLQEEPGA FNWSMYSQHA CLIEGKGECW QDKERQLRAR VKRVLPIDVH QPQPLGAGSP APLPADALVS AFCLEAVSPD LASFQRALDH ITTLLRPGGH LLLIGALEES WYLAGEARLT VVPVSEEEVR EALVRSGYKV RDLRTYIMPA HLQTGVDDVK GVFFAWAQKV GL | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The PNMT protein solution contains 20mM Tris-HCl, pH 8 and 10% Glycerol. |