missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPA1 Protein
A cDNA sequence encoding the PPA1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13113-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
PPA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSGFST EERAAPFSLE YRVFLKNEKG QYISPFHDIP IYADKDVFHM VVEVPRWSNA KMEIATKDPL NPIKQDVKKG KLRYVANLFP YKGYIWNYGA IPQTWEDPGH NDKHTGCCGD NDPIDVCEIG SKVCARGEII GVKVLGILAM IDEGETDWKV IAINVDDPDA ANYNDINDVK RLKPGYLEAT VDWFRRYKVP DGKPENEFAF NAEFKDKDFA IDIIKSTHDH WKALVTKKTN GKGISCMNTT LSESPFKCDP DAARAIVDAL PPPCESACTV PTDVDKWFHH QKN | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PPA1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol. |