missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRPS2 Protein
A cDNA sequence encoding the PRPS2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13170-5ug
Dettagli aggiuntivi : Netto-Gewicht : 0.01000kg
Specifica
PRPS2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPNIVLF SGSSHQDLSQ RVADRLGLEL GKVVTKKFSN QETSVEIGES VRGEDVYIIQ SGCGEINDNL MELLIMINAC KIASSSRVTA VIPCFPYARQ DKKDKVGESR APISAKLVAN MLSVAGADHI ITMDLHASQI QGFFDIPVDN LYAEPAVLQW IRENIAEWKN CIIVSPDAGG AKRVTSIADR LNVEFALIHK ERKKANEVDR MVLVGDVKDR VAILVDDMAD TCGTICHAAD KLLSAGATKV YAILTHGIFS GPAISRINNA AFEAVVVTNT IPQEDKMKHC TKIQVIDISM ILAEAIRRTH NGESVSYLFS HVPL | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRPS2 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |