missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAMP1 Protein
A cDNA sequence encoding the RAMP1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13259-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
RAMP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSCQEANYG ALLRELCLTQ FQVDMEAVGE TLWCDWGRTI RSYRELADCT WHMAEKLGCF WPNAEVDRFF LAVHGRYFRS CPISGRAVRD PPGS | |
Greater than 80% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAMP1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |