missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RHOB Protein
A cDNA sequence encoding the RHOB was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13314-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
RHOB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAIRKKLVV VGDGACGKTC LLIVFSKDEF PEVYVPTVFE NYVADIEVDG KQVELALWDT AGQEDYDRLR PLSYPDTDVI LMCFSVDSPD SLENIPEKWV PEVKHFCPNV PIILVANKKD LRSDEHVRTE LARMKQEPVR TDDGRAMAVR IQAYDYLECS AKTKEGVREV FETATRAALQ KRYGSQNGCI NCC | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RHOB protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 20% glycerol and 0.2mM NaCl. |