missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAR1B Protein
A cDNA sequence encoding the SAR1B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13414-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
SAR1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSFIFDW IYSGFSSVLQ FLGLYKKTGK LVFLGLDNAG KTTLLHMLKD DRLGQHVPTL HPTSEELTIA GMTFTTFDLG GHVQARRVWK NYLPAINGIV FLVDCADHER LLESKEELDS LMTDETIANV PILILGNKID RPEAISEERL REMFGLYGQT TGKGSISLKE LNARPLEVFM CSVLKRQGYG EGFRWMAQYI D | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SAR1B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |