Learn More
enQuireBio™ Recombinant Human SSR1 Protein
A cDNA sequence encoding the SSR1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13594-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
6745 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRGGPRGL LAVAQDLTED EETVEDSIIE DEDDEAEVEE DEPTDLVEDK EEEDVSGEPE ASPSADTTIL FVKGEDFPAN NIVKFLVGFT NKGTEDFIVE SLDASFRYPQ DYQFYIQNFT ALPLNTVVPP QRQATFEYSF IPAEPMGGRP FGLVINLNYK DLNGNVFQDA VFNQTVTVIE REDGLDGET | |
Greater than 90.0% as determined by SDS-PAGE. |
SSR1 Protein | |
Research Use Only | |
SSR1 | |
Human | |
His | |
SSR1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |