missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Steroid hormone receptor ERR1 Protein
A DNA sequence encoding the Homo sapiens (Human) Steroid hormone receptor ERR1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6001-ec-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001269379.1 | |
Steroid hormone receptor ERR1 Protein | |
Research Use Only | |
ESRRA | |
Recombinant Protein | |
E. coli | |
MSSQVVGIEP LYIKAEPASP DSPKGSSETE TEPPVALAPG PAPTRCLPGH KEEEDGEGAG PGEQGGGKLV LSSLPKRLCL VCGDVASGYH YGVASCEACK AFFKRTIQGS IEYSCPASNE CEITKRRRKA CQACRFTKCL RVGMLKEGVR LDRVRGGRQK YKRRPEVDPL PFPGPFPAGP LAVAGGPRKT AAPVNALVSH LLVVEPEKLY AMPDPAGPDG HLPAVATLCD LFDREIVVTI SWAKSIPGFS SLSLSDQMSV LQSVWMEVLV LGVAQRSLPL QDELAFAEDL VLDEEGARAA GLGELGAALL QLVRRLQALR LEREEYVLLK ALALANSDSV HIEDAEAVEQ LREALHEALL EYEAGRAGPG GGAERRRAGR LLLTLPLLRQ TAGKVLAHFY GVKLEGKVPM HKLFLEMLEA MMD | |
Greater than 80% as determined by SDS-PAGE. |
2101 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |