missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H1N1 Puerto Rico Recombinant Protein
A cDNA sequence encoding the H1N1 Puerto Rico Recombinant was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12077-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Influenza H1N1 Puerto Rico Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H1N1 A/Puerto Rico 08/1934 solution contains 10mM Sodium phosphate, pH 7.2, 150mM NaCl and 0.005% Tween-20. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |