missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RAET1E Protein
A cDNA sequence encoding the RAET1E was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13253-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
RAET1E Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
LDDAHSLRCN LTIKDPTSAD LPWCDVKCSV DEITILHLNN INKTMTSGDP GKMANATGKC LTQPLNDLCQ ELRDKVSNTK VDTHKTNGYP HLQVTMIYPQ SQGQTPSATW EFNISDSYFF TFYTENMSWR SANDESGVIM NKWKDDGDLV QQLKYFIPQC RQKIDEFLKQ SKEKPRSTSR SPSITQLTST SPLPPPSHSL EHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
RAET1E protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |